Learn More
LICOR IRDye 700 AP-1 Consensus Oligonucleotide (25 uL)
Catalog No. NC2209428
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. AP-1 is a transcription factor that regulates gene expression and controls a number of cellular processes including differentiation, proliferation, and apoptosis. For this reason, it is a common oligo used in EMSA to assess protein binding.
Catalog No. NC2209428 Supplier LICOR Supplier No. 82907925
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. AP-1 is a transcription factor that regulates gene expression and controls a number of cellular processes including differentiation, proliferation, and apoptosis. For this reason, it is a common oligo used in EMSA to assess protein binding.