Learn More
LICORbio IRDye 700 HIF-1 Consensus Oligonucleotide (25 uL)
Catalog No. NC2209430
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. HIF-1 is a transcription factor that regulates celluar responses to hypoxia, which have been heavily implicated in cancer biology and various other pathophysiologies. For this reason, it is a common oligonucleotide used in EMSA assays in basic discovery and therapeutics research.
This item is not returnable. View return policy
Catalog No. NC2209430 Supplier LICORbio Supplier No. 82907929
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. HIF-1 is a transcription factor that regulates celluar responses to hypoxia, which have been heavily implicated in cancer biology and various other pathophysiologies. For this reason, it is a common oligonucleotide used in EMSA assays in basic discovery and therapeutics research.