Learn More
LICORbio IRDye 700 NFB Consensus Oligonucleotide (25 L)
Catalog No. NC9927947
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. NFκB is a transcription factor that controls transcription of DNA, cytokine production, and cell survival. For this reason, it is a common oligonucleotide used in EMSA to assess the binding of specific proteins.
Catalog No. NC9927947 Supplier LICORbio Supplier No. 82907924
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. NFκB is a transcription factor that controls transcription of DNA, cytokine production, and cell survival. For this reason, it is a common oligonucleotide used in EMSA to assess the binding of specific proteins.