Learn More
LICORbio IRDye 700 Phosphoramidite (100 mol)
Catalog No. NC2209441
Encompass_Preferred
IRDye 700 Phosphoramidite has been optimized for the 700 nm channel of the LI-COR 4300 DNA Analysis System.
This item is not returnable. View return policy
Catalog No. NC2209441 Supplier LICORbio Supplier No. 420033
IRDye 700 Phosphoramidite has been optimized for the 700 nm channel of the LI-COR 4300 DNA Analysis System.