Learn More
LICOR IRDye 700 Sp-1 Consensus Oligonucleotide (25 uL)
Catalog No. NC2209429
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. Sp-1 is a transcription factor that regulates gene expression and controls a number of cellular processes including differentiation, proliferation, and apoptosis. For this reason, it is a common oligonucleotide used in EMSA to assess certain types protein binding.
This item is not returnable. View return policy
Catalog No. NC2209429 Supplier LICOR Supplier No. 82907926
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. Sp-1 is a transcription factor that regulates gene expression and controls a number of cellular processes including differentiation, proliferation, and apoptosis. For this reason, it is a common oligonucleotide used in EMSA to assess certain types protein binding.