Learn More
LICOR IRDye 700 STAT3 Consensus Oligonucleotide (25 uL)
Catalog No. NC2209418
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. STAT3 is a transcription factor that is activated by phosphorylation of tyrosine 705 and in response to particular ligands. For this reason, it is a common oligonucleotide used in EMSA to assess certain types of protein binding.
This item is not returnable. View return policy
Catalog No. NC2209418 Supplier LICOR Supplier No. 82907922
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. STAT3 is a transcription factor that is activated by phosphorylation of tyrosine 705 and in response to particular ligands. For this reason, it is a common oligonucleotide used in EMSA to assess certain types of protein binding.