Learn More
LICORbio IRDye 750 Maleimide Infrared Dye ( 0.5mg)
Catalog No. NC2216033
Encompass_Preferred
IRDye 750 Maleimide is an infrared dye with detection near 750 nm. Maleimide reactive group provides the functionality for labeling molecules that contain free sulfhydryl (–SH thiol) groups. This reactive group allows conjugation reactions to be performed at physiological pH. This dye is not suitable for use with LI-COR Odyssey Imaging Systems.
This item is not returnable. View return policy
Catalog No. NC2216033 Supplier LICORbio Supplier No. 92971040
IRDye 750 Maleimide is an infrared dye with detection near 750 nm. Maleimide reactive group provides the functionality for labeling molecules that contain free sulfhydryl (–SH thiol) groups. This reactive group allows conjugation reactions to be performed at physiological pH. This dye is not suitable for use with LI-COR Odyssey Imaging Systems.