Learn More
LICORbio IRDye 750 NHS Ester (0.5mg)
Catalog No. NC2209480
Encompass_Preferred
IRDye 750 NHS Ester is an infrared dye with detection near 750 nm. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins. This dye is not suitable for use with LI-COR Odyssey Imaging Systems.
This item is not returnable. View return policy
Catalog No. NC2209480 Supplier LICORbio Supplier No. 92970040
IRDye 750 NHS Ester is an infrared dye with detection near 750 nm. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins. This dye is not suitable for use with LI-COR Odyssey Imaging Systems.