Learn More
LICOR IRDye 750 NHS Ester (5mg)
Supplier:  LICOR 92970041
Encompass_Preferred
IRDye 750 NHS Ester is an infrared dye with detection near 750 nm. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins. This dye is not suitable for use with LI-COR Odyssey Imaging Systems.
Catalog No. NC2209481