Learn More
LICORbio IRDye 800 Phosphoramidite (100 mol)
Catalog No. NC2209440
Encompass_Preferred
IRDye 800 Phosphoramidite has been optimized for the 800 nm channel of the LI-COR 4300 DNA Analysis System.
This item is not returnable. View return policy
Catalog No. NC2209440 Supplier LICORbio Supplier No. 400033
IRDye 800 Phosphoramidite has been optimized for the 800 nm channel of the LI-COR 4300 DNA Analysis System.