Learn More
LICORbio IRDye 800CW Alkyne Infrared Dye (5mg)
Catalog No. NC2209459
Encompass_Preferred
IRDye 800CW Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 800CW.
This item is not returnable. View return policy
Catalog No. NC2209459 Supplier LICORbio Supplier No. 92965002
IRDye 800CW Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 800CW.