Learn More
LICORbio IRDye 800CW Azide Infrared Dye (50 mg)
Catalog No. NC1933752
Encompass_Preferred
IRDye 800CW Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.
Catalog No. NC1933752 Supplier LICORbio Supplier No. 92975050
IRDye 800CW Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.