Learn More
LICOR IRDye 800CW Azide Infrared Dye (0.5 mg)
Catalog No. NC0567231
Encompass_Preferred
IRDye 800CW Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.
Catalog No. NC0567231 Supplier LICOR Supplier No. 92960000
Add to Cart
Add to Cart
IRDye 800CW Azide provides the functionality for preferential labeling of molecules that contain either the alkyne or dibenzocyclooctyne (DBCO) group.