Learn More
LICORbio IRDye 800CW Carboxylate (500 mg)
Catalog No. NC2216027
Encompass_Preferred
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control. Note: The carboxylate dye has no reactive group and cannot be used for labeling.
This item is not returnable. View return policy
Catalog No. NC2216027 Supplier LICORbio Supplier No. 92970125
Add to Cart
Add to Cart
Request a Quote
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control. Note: The carboxylate dye has no reactive group and cannot be used for labeling.