Learn More
LICORbio IRDye 800CW Carboxylate (50 mg)
Catalog No. NC2209485
Encompass_Preferred
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control. Note: The carboxylate dye has no reactive group and cannot be used for labeling.
This item is not returnable. View return policy
Catalog No. NC2209485 Supplier LICORbio Supplier No. 92970122
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control. Note: The carboxylate dye has no reactive group and cannot be used for labeling.