Learn More
LICOR IRDye 800CW Carboxylate (5 mg)
Catalog No. NC0739341
Encompass_Preferred
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control.
Catalog No. NC0739341 Supplier LICOR Supplier No. 92909406
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control.