Learn More
LICORbio IRDye 800CW DBCO Infrared Dye (5 mg)
Catalog No. NC2209447
Encompass_Preferred
IRDye 800CW DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 800CW.
This item is not returnable. View return policy
Catalog No. NC2209447 Supplier LICORbio Supplier No. 92955000
IRDye 800CW DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 800CW.