Learn More
LICOR IRDye 800CW DBCO Infrared Dye (50 mg)
Catalog No. NC2209452
Encompass_Preferred
IRDye 800CW DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 800CW.
This item is not returnable. View return policy
Catalog No. NC2209452 Supplier LICOR Supplier No. 92955050
IRDye 800CW DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 800CW.