Learn More
LICORbio IRDye 680RD DBCO Infrared Dye (0.5mg)
Catalog No. NC2209446
Encompass_Preferred
IRDye 680RD DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 680RD.
This item is not returnable. View return policy
Catalog No. NC2209446 Supplier LICORbio Supplier No. 92950005
IRDye 680RD DBCO provides the functionality for preferential labeling of molecules that contain azide groups (–N3) with IRDye 680RD.