Learn More
LICOR IRDye 800CW Donkey anti-Goat IgG Secondary Antibody (0.5 mg)
Catalog No. NC9604570
Encompass_Preferred
Immunogen: Goat IgG, whole molecule | Applications: IRDye 800CW Donkey anti-Goat IgG Secondary Antibody is highly recommended for Western Blot, In-Cell Western™ Assay, On-Cell Western Assay, Protein Array, Immunohistochemistry, Small Animal Imaging, Microscopy, 2D Gel Detection, Tissue Section Imaging, and Virus Titration Assay. Note: Do not use this secondary antibody in combination with any secondary antibody whose host is goat (for example, IRDye 680RD Goat anti-Rabbit) for two-color Western blot detection. | Formulation: IRDye 800CW Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. NC9604570 Supplier LICOR Supplier No. 92632214
Immunogen: Goat IgG, whole molecule | Applications: IRDye 800CW Donkey anti-Goat IgG Secondary Antibody is highly recommended for Western Blot, In-Cell Western™ Assay, On-Cell Western Assay, Protein Array, Immunohistochemistry, Small Animal Imaging, Microscopy, 2D Gel Detection, Tissue Section Imaging, and Virus Titration Assay. Note: Do not use this secondary antibody in combination with any secondary antibody whose host is goat (for example, IRDye 680RD Goat anti-Rabbit) for two-color Western blot detection. | Formulation: IRDye 800CW Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.