Learn More
LICOR IRDye 800CW Maleimide Infrared Dye (5mg)
Catalog No. NC2216039
Encompass_Preferred
IRDye 800CW Maleimide provides the functionality for labeling molecules that contain free sulfhydryl (–SH) groups, such as cysteine residues in proteins, with IRDye 800CW. The reactive group of the IRDye 800CW Maleimide allows conjugation reactions to be performed very efficiently at physiological pH.
This item is not returnable. View return policy
Catalog No. NC2216039 Supplier LICOR Supplier No. 92980021
IRDye 800CW Maleimide provides the functionality for labeling molecules that contain free sulfhydryl (–SH) groups, such as cysteine residues in proteins, with IRDye 800CW. The reactive group of the IRDye 800CW Maleimide allows conjugation reactions to be performed very efficiently at physiological pH.