Learn More
LICORbio IRDye 800CW NHS Ester (500mg)
Catalog No. NC2209478
Encompass_Preferred
IRDye infrared dyes from LI-COR Biosciences are available for researchers developing applications using near-infrared (NIR) fluorescence. These unique dyes are used in a variety of NIR imaging applications including Western blotting, plate-based assays, protein arrays, tissue section imaging, and molecular activity measurements. Standard NHS ester chemistry is used to produce custom probes labeled with LI-COR IRDye infrared dyes. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins.
This item is not returnable. View return policy
Catalog No. NC2209478 Supplier LICORbio Supplier No. 92970025
Add to Cart
Add to Cart
Request a Quote
IRDye infrared dyes from LI-COR Biosciences are available for researchers developing applications using near-infrared (NIR) fluorescence. These unique dyes are used in a variety of NIR imaging applications including Western blotting, plate-based assays, protein arrays, tissue section imaging, and molecular activity measurements. Standard NHS ester chemistry is used to produce custom probes labeled with LI-COR IRDye infrared dyes. The NHS ester reactive group provides the functionality for labeling primary and secondary amines, such as lysine residues in proteins.