Learn More
Medchemexpress LLC Irdye 800CW NHS ester | 956579-01-4 | >80.0% | 10 MG
SDP
Catalog No. 5000298848
Encompass_Preferred
IRDye 800CW NHS ester is a near-infrared fluorescent dye supplied as an NHS ester for covalent labeling of primary and secondary amines, including lysine residues in proteins, peptides, and oligonucleotides. It is formulated for research use and is suitable for antibody and protein conjugation workflows.
  • Near-infrared fluorescent dye for low-background imaging and deep tissue detection.
  • NHS-ester reactive group enables covalent labeling of primary and secondary amines.
  • Absorption maxima ~774-778 nm and emission maxima ~789-794 nm for NIR applications.
  • High water solubility compatible with protein and peptide conjugation.
  • Available in multiple milligram sizes to accommodate small- and large-scale experiments.
Catalog No. 50-002-98848 Supplier Medchemexpress LLC Supplier No. HYD187210MG
IRDye 800CW NHS ester is a near-infrared fluorescent dye supplied as an NHS ester for covalent labeling of primary and secondary amines, including lysine residues in proteins, peptides, and oligonucleotides. It is formulated for research use and is suitable for antibody and protein conjugation workflows.
  • Near-infrared fluorescent dye for low-background imaging and deep tissue detection.
  • NHS-ester reactive group enables covalent labeling of primary and secondary amines.
  • Absorption maxima ~774-778 nm and emission maxima ~789-794 nm for NIR applications.
  • High water solubility compatible with protein and peptide conjugation.
  • Available in multiple milligram sizes to accommodate small- and large-scale experiments.