Learn More
LICORbio IRDye 800CW Protein Labeling Kits (microscale kit)
Catalog No. NC0273641
Encompass_Preferred
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays.The microscale kit is optimized for labeling 100 μg of protein with molecular weight 14 - 200 kDa. This kit contains 3 x 0.1 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin Column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - Microscale
Catalog No. NC0273641 Supplier LICORbio Supplier No. 92838044
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays.The microscale kit is optimized for labeling 100 μg of protein with molecular weight 14 - 200 kDa. This kit contains 3 x 0.1 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin Column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - Microscale