Learn More
LICORbio IRDye 800CW Protein Labeling Kits (low molecular weight kit)
Catalog No. NC2216041
Encompass_Preferred
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays. The low molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 14 - 45 kDa. This kit contains 3 x 0.5 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - Low MW
This item is not returnable. View return policy
Catalog No. NC2216041 Supplier LICORbio Supplier No. 92838042
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays. The low molecular weight kit is optimized for labeling 1.0 mg of protein with molecular weight 14 - 45 kDa. This kit contains 3 x 0.5 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - Low MW