Learn More
LICOR IRDye 800CW Protein Labeling Kits (high molecular weight kit)
Catalog No. NC9528510
Encompass_Preferred
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays. The high molecular weight kit is optimized for labeling 1.0 mg amounts of protein with molecular weight 45 - 200 kDa. This kit contains 3 x 0.1 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - High MW
Catalog No. NC9528510 Supplier LICOR Supplier No. 92838040
IRDye 800CW Protein Labeling Kits can be used to label antibodies and other proteins for applications such as Western blots, In-Cell Western™ assays, in vivo imaging, and whole organ or tissue section assays. The high molecular weight kit is optimized for labeling 1.0 mg amounts of protein with molecular weight 45 - 200 kDa. This kit contains 3 x 0.1 mg IRDye 800CW Reactive Dye vials, 0.5 mL 1 M Potassium Phosphate (K2HPO4) (pH 9), 25 mL 1X PBS, 0.5 mL ultra-pure water, 3 x Pierce Zeba™ Desalting Spin Columns, Pierce Zeba Desalting Spin column instructions, and Protocol for IRDye 800CW Protein Labeling Kit - High MW