Learn More
LICORbio IRDye 800CW Rabbit anti-HRP (0.5 mg)
Catalog No. NC1861701
Encompass_Preferred
Immunogen: Peroxidase from horseradish roots | Application: IRDye 800CW Rabbit anti-HRP is recommended for Western Blot | Formulation: IRDye 800CW secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. NC1861701 Supplier LICORbio Supplier No. 92632202
Immunogen: Peroxidase from horseradish roots | Application: IRDye 800CW Rabbit anti-HRP is recommended for Western Blot | Formulation: IRDye 800CW secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.