Learn More
LICOR IRDye 800RS NHS Ester (0.5 mg)
Catalog No. NC1746573
Encompass_Preferred
IRDye 800RS infrared dye is an excellent choice for nucleic acid applications. This near-infrared fluorescence dye has good water solubility, but low salt tolerance. It is more hydrophobic than IRDye 800CW. IRDye 800RS NHS ester can be used to label the primary and secondary amino groups of DNA and RNA. Nucleic acids that have been labeled with IRDye 800RS can be easily purified by reverse-phase chromatography.
Catalog No. NC1746573 Supplier LICOR Supplier No. 92972020
IRDye 800RS infrared dye is an excellent choice for nucleic acid applications. This near-infrared fluorescence dye has good water solubility, but low salt tolerance. It is more hydrophobic than IRDye 800CW. IRDye 800RS NHS ester can be used to label the primary and secondary amino groups of DNA and RNA. Nucleic acids that have been labeled with IRDye 800RS can be easily purified by reverse-phase chromatography.