Learn More
LICORbio IRDye QC-1 NHS Ester (0.5 mg)
Catalog No. NC0625367
Encompass_Preferred
IRDye QC-1 is the first non-fluorescent (dark) quencher compatible with a wide range of visible and near-infrared fluorophores (~500-800 nm). It has the widest available quenching range, so you do not need to carefully match the donor's fluorescence spectrum with the acceptor's absorbance. IRDye QC-1 NHS ester can be used for FRET (fluorescence resonance energy transfer) applications such as protease assays. Recently, it has been paired with IRDye 800CW in optoacoustic studies.
Catalog No. NC0625367 Supplier LICORbio Supplier No. 92970030
IRDye QC-1 is the first non-fluorescent (dark) quencher compatible with a wide range of visible and near-infrared fluorophores (~500-800 nm). It has the widest available quenching range, so you do not need to carefully match the donor's fluorescence spectrum with the acceptor's absorbance. IRDye QC-1 NHS ester can be used for FRET (fluorescence resonance energy transfer) applications such as protease assays. Recently, it has been paired with IRDye 800CW in optoacoustic studies.