Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Monoclonal Antibody, IRF-8, Rabbit, Human, 100 μL
SDP

Catalog No. 502730868
Encompass

This rabbit monoclonal antibody is designed to recognize endogenous levels of total IRF-8 protein. Produced by immunizing animals with a recombinant protein specific to the amino terminus of human IRF-8, this primary antibody is validated for various applications including Western blotting, immunoprecipitation, immunohistochemistry, and chromatin immunoprecipitation.

  • Recognizes endogenous levels of total IRF-8 protein
  • Validated for multiple applications including Western blotting, ChIP, and IHC
  • Host species: rabbit
  • Species reactivity: human
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide

Catalog No. 50-273-0868 Supplier Cell Signaling Technology Supplier No. 83413S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody is designed to recognize endogenous levels of total IRF-8 protein. Produced by immunizing animals with a recombinant protein specific to the amino terminus of human IRF-8, this primary antibody is validated for various applications including Western blotting, immunoprecipitation, immunohistochemistry, and chromatin immunoprecipitation.

  • Recognizes endogenous levels of total IRF-8 protein
  • Validated for multiple applications including Western blotting, ChIP, and IHC
  • Host species: rabbit
  • Species reactivity: human
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.