Learn More
Aviva Systems Biology Corp IRF1 Antibody - N-terminal region (ARP31296_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31296_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRF1. Alternate names: IRF-1, MARApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54193