Learn More
Aviva Systems Biology Corp Irf1 Antibody - middle region (ARP36903_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36903_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Irf1. It was validated on Western Blot. Irf1,interferon regulatory factor 1,Irf-1; AU020929; Irf1,interferon regulatory factor 1,interferon regulatory factor 1,OTTMUSP00000005820,OTTMUSP00000005821,OTTMUSP00000036703The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AU020929, Irf-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-284-540