Learn More
Aviva Systems Biology Corp IRF2 Antibody - N-terminal region (P100943_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100943_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRF2. Alternate names: IRF-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-62793