Learn More
Aviva Systems Biology Corp IRF2 Antibody - N-terminal region (P100613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRF2. Alternate names: IRF-2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-014-62664