Learn More
Aviva Systems Biology Corp IRF3 Antibody - C-terminal region (ARP31992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human IRF3. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54409