Learn More
Cayman Chemical IRF3S386AS396A mutnthu 100ug
SDP
Catalog No. 502995289
Encompass_Preferred
Source: N-terminal His-tagged human IRF3 (S386A, S396A mutant) purified from E. coli • MW: 47.2 kDa
Catalog No. 50-299-5289 Supplier Cayman Chemical Supplier No. 23590100 UG
Source: N-terminal His-tagged human IRF3 (S386A, S396A mutant) purified from E. coli • MW: 47.2 kDa