Learn More
Aviva Systems Biology Corp IRF4 Antibody - middle region (ARP38155_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38155_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IRF4. Alternate names: LSIRF, MUM1, NF-EM5ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-56542