Learn More
Aviva Systems Biology Corp IRF4 Antibody - N-terminal region (ARP38154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IRF4. Alternate names: LSIRF, MUM1, NF-EM5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56541