Learn More
Cayman Chemical IRF4MUM1CTerm RabbIt Mon 100uL
SDP
Catalog No. 502948566
Encompass_Preferred
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal
Catalog No. 50-294-8566 Supplier Cayman Chemical Supplier No. 32282100 UL
A catechol estrogen and minor metabolite of estrogen production is increased in microsomes prepared from human mammary adenocarcinoma and fibroadenoma microsomes compared with microsomes prepared from non-cancerous tissues induces renal tumor formation in male hamsters at 25 mg/animal