Learn More
Aviva Systems Biology Corp IRF5 Antibody - N-terminal region (ARP38131_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38131_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRF5. Alternate names: SLEB10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56528