Learn More
Aviva Systems Biology Corp IRF5 Antibody - N-terminal region (ARP37180_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37180_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse IRF5. Alternate names: mirf5, AW491843ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56333