Learn More
Invivogen THP1-Dual™ KO-IRF5 Cells, IRF5 Knockout, Monocytic Cell Line, 2 Cells
SDP
Supplier:  Invivogen THPDKOIRF5
Encompass
THP1-Dual™ KO-IRF5 Cells provide a robust platform for studying the role of IRF5 in immune signaling. Engineered from THP1-Dual™ cells with a stable knockout of the IRF5 gene, they enable simultaneous monitoring of IRF and NF-κB pathways via Lucia luciferase and SEAP reporter genes. Functional validation confirms reduced NF-κB responses upon stimulation, making these cells essential for research into IRF5-targeting therapeutics.
Catalog No. NC2057746