Learn More
Aviva Systems Biology Corp IRF6 Antibody - N-terminal region (ARP38738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IRF6. Alternate names: LPS, OFC6, PIT, PPS, VWS, VWS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56775