Learn More
Aviva Systems Biology Corp IRF7 Antibody - middle region (ARP38415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IRF7. Alternate names: IRF-7H, IRF7AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56645