Learn More
Aviva Systems Biology Corp IRF8 Antibody - N-terminal region (P100841_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100841_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRF8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human IRF8. Alternate names: ICSBP, IRF-8, ICSBP1, H-ICSBPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53803