Learn More
Medchemexpress LLC Irganox 1330 | 1709-70-2 | 10 G
SDP
Catalog No. 5000234221
Encompass_Preferred
Irganox 1330 is a sterically hindered phenolic antioxidant used to protect polymers from thermal and oxidative degradation during processing and service. It is commonly applied in polyolefins to improve thermal stability, color retention, and resistance to extraction.
  • Protects polymers from oxidation during processing and use.
  • Improves thermal stability of polyolefins.
  • Reduces color formation and preserves appearance.
  • Exhibits low volatility and good extraction resistance.
  • Compatible with polyethylene, polypropylene, and other plastics.
Catalog No. 50-002-34221 Supplier Medchemexpress LLC Supplier No. HY12038810G
Irganox 1330 is a sterically hindered phenolic antioxidant used to protect polymers from thermal and oxidative degradation during processing and service. It is commonly applied in polyolefins to improve thermal stability, color retention, and resistance to extraction.
  • Protects polymers from oxidation during processing and use.
  • Improves thermal stability of polyolefins.
  • Reduces color formation and preserves appearance.
  • Exhibits low volatility and good extraction resistance.
  • Compatible with polyethylene, polypropylene, and other plastics.