Learn More
Irgasan (Triclosan), G-Biosciences

Description
It is often used as a supplement to cell culture media in Pseudomonas and Yersinia isolation agars to prevent bacterial contamination. It is slightly soluble in water and freely soluble in ethanol and methanol.
Specifications
Specifications
| Form | Powder |
| Product Line | Gibco |
| Purity or Quality Grade | Molecular Biology |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 289.54 |
| Purity | >98% |
| Boiling Point | 120°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.