Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci IRGB10 (CT) Peptide
SDP

Catalog No. 16595718
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-718 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-718 Supplier ProSci Supplier No. 9515P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

IRGB10 (CT) Peptideis used for blocking the activity of theIRGB10 (CT) Antibody

IRGB10 Antibody: IRGB10 is an inferno-inducible protein that is essential for the activation of DNA-sensing AIM2 inflammasome, and contributes to the activation of LPS-sensing caspase-11 and NLRP3 inflammasome. IRGB10 is part of a conserved signaling hub at the interface between cell-autonomous immunity and innate immune sensing pathways. IRGB10 targets cytoplasmic bacteria and compromises bacterial structural integrity. IRGB10 and LIGP2 act together to confer differential susceptibility against Chlamydia infection in mice.

Specifications

Gene ID (Entrez) 631323
Species Mouse
Accession Number AGY29631
Common Name predicted gene 12250
Concentration 200 μg/mL
Content And Storage Store IRGB10 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol IRGB10
Quantity 0.05 mg
Sequence 13 amino acids near the carboxy terminus of IRGB10 (CT).
Synonym IRGB10 Antibody: Immunity-related GTPase family member b10, Interferon-gamma-inducible p47 GTPase, Gm12250, OTTMUSG00000005723
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.