Learn More
Medchemexpress LLC HY-P0122 1mg Medchemexpress, iRGD peptide CAS:1392278-76-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP01221MG
Encompass_Preferred
Medchemexpress, HY-P0122 1mg iRGD peptide CAS:1392278-76-0 iRGD peptide is a 9-amino acid cyclic peptide, triggers tissue penetration of drugs by first binding to av integrins , then proteolytically cleaved in the tumor to produce CRGDK/R to interact with neuropilin-1, and has tumor-targeting and tumor-penetrating properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4025