Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium ICP Standard, 1000 ppm Ir in 10% HCl, Ricca Chemical
SDP

Catalog No. PIR1KH500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PIR1KH-500 500 mL Natural Poly Bottle
PIR1KH-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIR1KH-500 Supplier Ricca Chemical Company Supplier No. PIR1KH500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13569-57-8 , 7647-01-0 , 13569-57-8 , 7732-18-5
Molecular Formula Cl3H6IrO3
Molecular Weight (g/mol) 352.612
InChI Key LNJXVUXPFZKMNF-UHFFFAOYSA-K
PubChem CID 6102153
IUPAC Name trichloroiridium;trihydrate
SMILES O.O.O.Cl[Ir](Cl)Cl

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Ir
CAS Min % 88.0, 10.33, 0.16
CAS Max % 91.0, 10.75, 0.17
Equivalence 1 mL = 1 mg Ir
Physical Form Liquid
Color Brown
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Chemical Name or Material Iridium ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.